DXR Microscope stage drift in Z direction
Issue
During data collection the sample will drift out of focus.
Environment
- DXR Microscope
Resolution
- The stage Z mechanism needs to have the tension adjusted to hold the stage at the set focus.
- The attached document shows the knob that is used to increase the tension on the mechanism
Attachment(s)
| File | Last Modified |
|---|---|
| TIGHTEN Z AXIS drift.pdf | September 14, 2022 |