Location of Performance Verification files for OMNICxi
Issue
Where are the performance verification files for OMNICxi located?
Environment
- Raman DXRxi
- OMNICxi
Resolution
The test data associated with Performance Verification (PV) is located in C:\ProgramData\Thermo\OMNICxi\PV Reports.
The Program Data folder is a hidden folder. The folder options will need to be set to view hidden folders so you can drill down to the data.