Performance Test not included with OMNIC for Dispersive Raman
Issue
OMNIC For Dispersive Raman 9.4 no longer contains the dxrperftest on the OMNIC DVD.
Environment
OMNIC for Dispersive Raman 9.4
Resolution
- Download this file from the PSE website: OFDR 9.X DXR Perf Test Service Only.zip.
- Unzip the files on a flash drive or make a DVD.
- Run the tests from your flash drive, or DVD by double clicking on the executable. (Do not load the files onto the customer’s PC.)