AN 9146 - Determination of Hydroxyl Number in Polyols by Mid-infrared Spectroscopy
Description:
Quantitative infrared analysis of complex samples is often best accomplished using multivariate calibration techniques such as partial least squares (PLS).
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
AN 9146 - Determination of Hydroxyl Number in Polyols by Mid-infrared Spectroscopy.pdf | August 03, 2022 |