AN 9584 - Applications of Rapid Scan Data Collection
Description:
Rapid scan techniques allow one to record infrared spectra quickly and accurately in kinetic events such as photochemical reactions, temperature or pressure jump, flowing liquids or diffusion kinetics, fast reactions between ions and oxidation reduction processes.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
AN 9584 - Applications of Rapid Scan Data Collection.pdf | August 04, 2022 |