TN 9364 - Measuring the Signal-to-Noise Ratio of an Infrared Spectrometer
Description:
Evaluating the performance of an infrared spectrometer includes a number of tests such as measuring the signal-to-noise ratio (SNR), stability, spectral range, linearity, resolution, and wave length accuracy. While all of these tests are very important, the one which receives most frequent review is the signal-to-noise ratio.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
TN 9364 - Measuring the Signal-to-Noise Ratio of an Infrared Spectrometer.pdf | August 03, 2022 |