TN 9476 - FT-IR Sampling Parameters for Exhaust Gas Measurements
Description:
Fourier transform infrared (FT-IR)spectroscopy is a useful solution to the analytical problems of exhaust measure-ment. While traditional on-line analyzers require exhaust to be diluted and cooled before sampling, FT-IR can measure the raw exhaust directly. Many highly reactive species in exhaust can be measured by FT-IR down to low part-per-million levels.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
TN 9476 - FT-IR Sampling Parameters for Exhaust Gas Measurements.pdf | August 03, 2022 |