TN 9582 - Performance Comparison for Tri-Glycine Sulfate and Lithium Tantalate Detectors
Description:
Most mid infrared analysis is accomplished with a tri-glycine sulfate (TGS) detector.This detector remains the detector of choice due to its excellent linearity, ease of use, and good sensitivity.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
TN 9582 - Performance Comparison for Tri-Glycine Sulfate and Lithium Tantalate Detectors.pdf | August 04, 2022 |