Nicolet FTIR Beam Information
Issue
What is the aperture size of the Summit and the diameter of the beam?
Environment
- Nicolet Summit
- Nicolet Summit PRO
Resolution
Aperture sizes and beam diameters appear in the table below. Refer to the attachment for more information.
Instrument Model | Aperture | Aperture Size (mm) | IR Beam Diameter Sample Compartment (mm) | IR Beam Diameter At Detector (mm) |
Summit | Fixed Aperture | 1.85 | 5.78 | 1.16 |
Summit PRO | Large Aperture | 2.29 | 7.15 | 1.43 |
Summit PRO | Small Aperture | 0.81 | 2.53 | 0.51 |
Attachment(s)
File | Last Modified |
---|---|
Nicolet Summit FTIR Beam Information.pdf | December 16, 2021 |