Thermo Fisher Scientific
Unknown error, error number -2146893813
Issue
Unknown error, Error number -2146893813 is generated during an archive event
Environment
  • RESULT
  • Logged in as a Local User in the administrative group.
Resolution
Delete the folder: “C:\Users\username\AppData\Roaming\Microsoft\Crypto\RSA”