213-0029-01 - TempSet Instruction Card
Description:
The TempSet™program can be used as a troubleshooting tool to verify USB communication between the computer and the USB controller,and for manually setting the temperature of an accessory. This software does not require OMNIC software to be running.
Environment:
- FT-IR
- OMNIC
- TempSet
Attachment(s):
File | Last Modified |
---|---|
213-0029-01 - TempSet Instruction Card.pdf | August 04, 2022 |