269-176702 - Rev A - Polystyrene Reflection Standard for ATR Accessory Qualification
Description:
This document describes the traceable polystyrene standard for the attenuated total reflection (ATR) Smart accessories from Thermo Fisher Scientific. The standard can be used to verify the performance of a Thermo Scientific FT-IR spectrometer with the installed accessory.
The ATR standard is available for Thermo Scientific ATR Smart accessories purchased with the ValPro™ System Qualification package. For convenience, the standard is mounted in a holder designed for ATR sampling using the accessory’s built-in pressure device.
The ATR standard is traceable to a certified standard stored at our factory. This document explains the company’s internal process for measuring the ATR standard to establish traceability, including all associated data and reports.
Environment:
Thermo Scientific ATR Smart accessories purchased with the ValPro™ System Qualification package
Attachment(s):
File | Last Modified |
---|---|
269-176702 - Rev A - Polystyrene Reflection Standard for ATR Accessory Qualification.pdf | December 14, 2021 |
269-175202 - Rev A - Transmission Standards for FT-IR Spectrometer Qualification.pdf | December 14, 2021 |