Thermo Fisher Scientific
2I-15011 - Issue 13 - Atlas Manual 15Ton (15T) and 25Ton - User Manual
Description: 
Thank you for purchasing a Specac product.
The Specac Atlas™ Manual 15T and 25T Hydraulic Presses have been designed for a wide variety of pressing applications, but they are specifically suited to the preparation of KBr discs using Specac evacuable pellet die assemblies.
Environment:
Atlas Manual 15Ton (15T) and 25Ton