Thermo Fisher Scientific
DiffusIR - Product Data Sheet
Description: 
The PIKE Technologies environmental chambers can be operated at temperatures ranging from -150 to 900 ºC and at pressures up to 1500 psi. The optional heat chambers are easily inserted and removed from the DiffusIR using push lock pins.
Environment:
DiffusIR
  • Was this article helpful?