FT-IR - Troubleshooting Spectral Results
Description:
Specular reflectance experiments are non-destructive, easy, fast, require minimal sample preparation, and are quantifiable. This experiment is a surface technique, as this diagram suggests. The one requirement for successful collection is a flat, level surface. Inconsistencies in the surface structure will cause the energy to scatter which may make the interpretation of the data more difficult if not impossible.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
FT-IR - Troubleshooting Spectral Results.pdf | August 04, 2022 |