Multiple Reflection HATR - Product Data Sheet
Description:
Horizontal Attenuated Total Reflectance (HATR) accessories successfully replace constant path transmission cells, salt plates and KBr pellets used in the analysis of liquid, semi-liquid materials and a number of solids. HATRs feature a constant and reproducible effective pathlength and are well suited for both qualitative and quantitative applications. In general, sampling is achieved by placing the sample onto the HATR crystal – generally eliminating sample preparation.
Environment:
Multiple Reflection HATR
Attachment(s):
File | Last Modified |
---|---|
Multiple Reflection HATR - Product Data Sheet.pdf | August 04, 2022 |