Thermo Fisher Scientific
Peltier Liquid ATR Tower - Specification Sheet
Description: 
The Simplex Scientific Peltier Liquid ATR Tower package is designed to integrate temperature control to the Smart or Foundation Series Performer accessory that is equipped with a single bounce diamond crystal puck. Temperature can be manually set from the computer using TempSet™ software program or ramping and series experiments can be controlled from an optional software packages. Simplex Scientific TempProfile™ or @Temp™ software packages integrates with Thermo Scientific OMNIC software, allowing temperature controlled data collection. Connection to the PC is through the standard 2.0 USB (Universal Serial Bus). The Peltier Liquid ATR Tower has a temperature range of 6° C to 80° C.
Environment:
Peltier Liquid ATR Tower
  • Was this article helpful?