Thermo Electron - Spectral Libraries and Printed Books
Description:
Thermo Electron has over 81,000 high-quality FT-IR and Raman spectra available for use with Thermo FT-IR products. These libraries aid in FT-IR spectral analysis by providing a reference spectra with which to compare a sample spectrum. This is useful for identification of unknowns and verification of materials. The collection includes spectral references of general materials such as organic chemicals as well as a variety of industry-specific libraries. A selection of reference publications of general interest to the infrared or Raman spectroscopist is also available.
Environment:
Thermo Electron
Attachment(s):
File | Last Modified |
---|---|
Thermo Electron - Spectral Libraries and Printed Books.pdf | August 05, 2022 |