VeeMAX III with ATR Variable Angle Single Reflection ATR for Depth Profiling Studies
Description:
The VeeMAX III with ATR offers continuous variable angle of incidence and a variety of crystal plates to selectively control the depth of penetration of the IR beam into the sample. ATR applications include the study of layered samples, coatings, release agents, monolayers on silicon and chemical migration studies.
Environment:
VeeMAX III
FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| VeeMAX III with ATR - Variable Angle, Single Reflection ATR for Depth Profiling Studies.pdf | August 04, 2022 |