Replacing a Crystal Plate
Environment:
Smart ATR
Resolution:
To replace the crystal plate:
- Remove the two Phillips or straight blade screws securing the pressure device (circled in red below) and remove the pressure device.
- Remove the old plate and install the new plate.
- Carefully replace the pressure device and secure with the two screws.