AN 50813 - FT-NIR Analysis of Wine
Description:
This report demonstrates that the Nicolet Antaris FT-NIR analyzer can be used successfully for the simultaneous quantitative analysis of multiple components of wine. NIR can predict physical properties like density in addition to normal chemical parameters like ethanol content or Degrees Brix.
Environment:
Antaris FT-NIR
Attachment(s):
File | Last Modified |
---|---|
AN 50813 - FT-NIR Analysis of Wine.pdf | July 25, 2022 |