AN 51954 Rapid Multi-Component Analysis of Soybeans by FT-NIR Spectroscopy
Description:
This application study will show how the ThermoScientific Antaris Fourier transform near-infrared (FT-NIR) analyzer with the Sample Cup Spinner accessory (Figure 1) was successfully used to analyze whole soybeans. The calibration models developed during this study prove thatrapid, non-destructive near-infrared technology can accurately predict important components in soybeans.
Environment:
- FT-NIR
- NIR
- Antaris
Attachment(s):
File | Last Modified |
---|---|
AN 51954 - Rapid, Multi-Component Analysis of Soybeans by FT-NIR Spectroscopy.pdf | July 28, 2022 |