Differences in NIR Fiber Optic cables
Description:
Not all fiber optic cables are functional in NIR. We need to use low O-H fiber cables. Attached are some spectral response curves for various fiber optic cables.
Environment:
- Antaris II
- Antaris MX
- Antaris EX
Attachment(s):
File | Last Modified |
---|---|
Fiber Optic energy curves.pdf | July 07, 2022 |