Thermo Fisher Scientific
ISN 0007 - Quantitative Characterization of Lactose Crystalline Forms, A Case Study to Illustrate the Utility of FT-NIR as a Tool for the Analysis of the Crystalline Composition of Solid Materials
Description: 
In this study, FT-NIR was used to quantitatively characterize binary and ternary mixtures of lactose, an important excipient used in many types of pharmaceutical dosage forms.