TN 51241 Process data communications with the Antaris FT-NIR Analyzers
Description:
Using analytical data to improve manufacturing processes has been standard practice in many industries for decades. How the data is communicated and utilized depends greatly on the type of measurement that is needed. Some discrete, in-process measurements like temperature or pressure only require analog input or output whereas other, more complex, types of information like spectroscopic data sets require higher order data processing and communication. RESULT software provides a user-friendly environment for implementing various customizable communication protocols within a process environment.
Environment:
- Antaris FT-NIR Analyzers (Analog, Digital, OPC and LIMS)
- RESULT
Attachment(s):
File | Last Modified |
---|---|
Technical Note 51241 - Process Data Communications with the Antaris FT-NIR Analyzers – Analog, Digital, OPC and LIMS.pdf | July 12, 2022 |