Nicolet iN5 - Objective Specification Sheet
Description:
The objective on the iN5 microscope has a 10X magnification and numerical aperture of 0.71. The minimum incident angle is 19.4 degrees and the maximum angle is 46.0 degrees. The focal length is approximately 7.37 mm.
Environment:
Nicolet iN5
Attachment(s):
File | Last Modified |
---|---|
Nicolet iN5 - Specification Sheet.pdf | August 03, 2022 |