Connections and ports Last updated Save as PDF A USB 3.0 connection to computer's USB 3.0 port B Connection to the optional joystick C Connect to Auxiliary Signals port on the spectrometer D Connect to "Accessory" port on the spectrometer E Connection to power supply cable