You can use the optional joystick to control the stage position and sample illumination. The stage and illumination can also be controlled in the software.
To connect the joystick, plug the data cable into the "Joystick" port on the back of the microscope.
A
Transmittance illumination control
Rotate to manually dim or brighten transmittance illumination.
B
Reflectance illumination control
Rotate to manually dim or brighten reflectance illumination.
C
Joystick for stage control
Push the joystick forward, backward, left, or right to move the stage along the sample plane.
Rotate to move the stage up or down.
D
Speed control
Controls the speed of the stage for precise, slow movement or fast movement.