Lifting or moving the instrument Last updated Save as PDF This instrument weighs approximately 70.3 kg (155 lb) and should be lifted by at least two people. To avoid risk of injury, use proper lifting techniques.