Thermo Fisher Scientific
Utility requirements
If at all possible, the power connections for the instrument and accessories should be easily accessible for service purposes.
Any line for dry air or nitrogen, which is used to purge the system, should also be accessible if service is required, and you should have direct control over the system utilities.
It is important to have all system utilities installed before the instrument arrives. Utility installations must comply with all local building and safety codes.
  • Was this article helpful?