Thermo Fisher Scientific
Optional Trinocular Eyepiece
Your microscope may have either a monocular eyepiece with only the camera or it may have the trinocular eyepiece with the camera and visual eyepieces.
raptir optional eyepieces.jpg
A Visual eyepieces Adjustable eyepieces for looking at the sample. Best used with the optional joystick.
B Three-position view selector
Controls the light path for the eyepieces.
  • In: Eyepiece only, no camera
  • Middle: Eyepiece and camera
  • Out: Camera only, no eyepieces
C Camera The USB camera is operated with OMNIC Paradigm software.
  • Was this article helpful?