Thermo Fisher Scientific
About the warranty
Your spectrometer is designed to work reliably for many years, and our software features allow you to keep careful track of its performance. We stand behind our instruments by providing a warranty for the entire system for 12 months (14 months from shipment date).
Your warranty does not cover damage to hygroscopic parts (the purge windows and the beamsplitter) if the spectrometer is exposed to excessive moisture, or if the cover is removed for an extended time by someone other than one of our representatives. To see the complete warranty or warranties for your spectrometer, see the documentation set for the system.
  • Was this article helpful?