Thermo Fisher Scientific
What's in the box
Your spectrometer will be unpacked and installed by one of our service representatives.
Your spectrometer is intended to be unpacked and installed by one of our service representatives, but if necessary, you can unpack the shipping box before the installation.
To avoid permanent damage to the optical components in your spectrometer, do not open anything, especially the plastic bag that protects the spectrometer, until the entire shipping box has come to room temperature. Damage due to unpacking the spectrometer before it has come to room temperature is not covered by the warranty.
  • Was this article helpful?