Smart iTR - Specification Sheet
Description:
The Thermo Scientific Smart iTR™ is an ultra-high-performance, versatile Attenuated Total Reflectance (ATR) sampling accessory. Primarily designed for use with a single bounce diamond crystal, the Smart iTR also offers other options for full configurability. Each of the ATR crystals is designed for maximum infrared throughput and spectroscopic performance.
Environment:
Smart iTR
Attachment(s):
File | Last Modified |
---|---|
Smart iTR - Specification Sheet.pdf | August 02, 2022 |