Thermo Fisher Scientific
iD1H Heated Transmission Accessory - iD1H Brochure
Description: 
The Thermo Scientific™ iD1H Heated Transmission accessory is ideal for the Thermo Scientific™ Nicolet™ iS™5N FT-NIR spectrometer system, providing consistent temperature control of vials and cuvettes for precise quantitative liquid analysis. The self-adjusting sample holder and fixed temperature heating element provide flexibility and consistency to meet your NIR quantification needs. The iD1H Heated Transmission accessory offers many benefits:
• Spring-loaded sample holder for use with multiple vials and cuvettes
• Fixed 40 °C temperature control
• Embedded LED for visual indication of temperature stability
• Internal aperture for best resolution and line shapes
Environment:
iD1H Heated Transmission Accessory
  • Was this article helpful?