Thermo Fisher Scientific
Basic Omnic Specta Lab
Description: 
Omnic Specta is a new program with Omnic 8.0 that a customer may purchase as an option. Omnic Specta indexes or alphabetizes all spectra on the hard drive. This includes Spectra (.spa), Spectral groups (.spg) and all Omnic and user libraries defined in the configuration. This makes spectra manipulation very fast. The key feature of Omnic Specta is to make it easier for users to perform operations by having to click less to perform a function
Environment:
Omnic
  • Was this article helpful?