Thermo Fisher Scientific
Blue Touch-Point Setup
Description: 
The blue “touch point” button located on the iS50 interface for iN5 is one in a series of touch points on the iS50 depending on the iS50 configuration. Touch points serve as a quick and easy way to select a specific beampath, load the related default experiment file and start a data collection. The attached procedure describes how to set up the touch point button.
Environment:
  • Nicolet iN5
  • Nicolet iS50
  • Was this article helpful?