How To Run a Bench Status Report Nicolet iS50 and iS5
Description:
The attached document describes how to generate a bench status report saved to the workstation desktop..
Environment:
- Nicolet iS5
- Nicolet iS50
- OMNIC Software version 9 or higher
Resolution :
How to Run A Bench Status Report on an iS5 or iS50 FT-IR Bench with Omnic 9 Version Software
To generate a Bench Status Report for an iS5 or iS50 bench in the Omnic 9 version software or higher in the Omnic go to the Collect > Advanced Diagnostics.
The following screen shot appears:
On your keyboard hold down the Shift button while simultaneously left mouse clicking on the Report button above. The following message will appear:
Hit the OK button to send the bench status report to the desktop of your computer.
Service Personal may request this report for diagnostics and troubleshooting purposes.
Attachment(s):
File | Last Modified |
---|---|
Picture1.png | April 14, 2023 |
Picture2.png | April 14, 2023 |
Picture3.png | April 14, 2023 |