Nicolet iS50 - iS50 Built-in ATR Serial Number Conventions
Description:
As of September 2018, the iS50 built-in ATR has the serial number convention of AV_:
• AVP = crystal
• AVL = upper optics
• AVM = detector preamp
• AVK = flipper
• AVN = tower
Environment:
Nicolet iS50
Attachment(s):
File | Last Modified |
---|---|
Nicolet iS50 - iS50 Built-in ATR Serial Number Conventions.pdf | August 01, 2022 |