Thermo Fisher Scientific
Autosampler File Error
Issue
When using TGA Autosampler (jstarter) application after setting the parameters in the sample tab and trying to start the analysis, this error message appears:
Unable to allocate a file for series collection.
Environment
OMNIC
Resolution
Updating the filename corrects the issue.
Cause
The issue was found to be an invalid file name character.
  • Was this article helpful?