Thermo Fisher Scientific
AN 50581 - Advanced ATR Correction Algorithm
Description: 
This application note discusses the use of the Advanced ATR Correction algorithm to correct ATR collected spectra for known band intensity distortion, peak shifts, and non-polarization effects.
Environment:
OMNIC
Attenuated Total Reflection technique
  • Was this article helpful?