AN 9475 - Fiber Optic Sampling in the Near-infrared Region Utilizing a High-Performance Infrared Spectrometer
Description:
Near-infrared spectroscopy is a popular technique for many analytical applications for two main reasons: the sampling advantages of near-infrared over mid-infrared spectroscopy for certain types of organic materials and the availability of sophisticated chemometric software for composition and property determination.
Environment:
FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| AN 9475 - Fiber Optic Sampling in the Near-infrared Region Utilizing a High-Performance Infrared Spectrometer.pdf | August 04, 2022 |