AN 9585 - Infrared Microspectroscopy of Hard Samples Utilizing A Diamond ATR Objective
Description:
The use of attenuated total reflectance (ATR) microscopy has made infrared spectroscopic sample handling faster and easier. By simply bringing the ATR objective into contact with the sample and aperturing to the desired area, we can quickly generate high quality spectra of a wide variety of samples.
Environment:
FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| 9585 - FT-IR - Application Note.pdf | August 04, 2022 |