AN 9587 - Curve Fitting Techniques for FT-IR Spectra of Proteins
Description:
Infrared spectroscopists often face complex multi-component bands which overlap into a broad unresolved absorption. The individual component absorptions from condensed phase spectra which are hidden within a broad band cannot be resolved by increasing spectral resolution.
Environment:
FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| AN 9587 - Curve Fitting Techniques for FT-IR Spectra of Proteins.pdf | August 04, 2022 |