Thermo Fisher Scientific
The Use of a Mid-IR Fiber Optic Probe in Sugar Analysis
Description: 
The use of FT-IR spectroscopy for the determination of the sucrose contentin sugar cane juice has been demonstrated (1). This important analysis isoften used as a basis for cane sugar payment in the sugar cane industry.
Environment:
FT-IR
  • Was this article helpful?