Bench settings for using an InGaAs detector
Issue
What are the bench settings for using an InGaAs detector?
Environment
iS50 main bench
Resolution
When using an InGaAs detector in the main bench of an iS50, it is important to set the correct parameters on the Bench tab of Experiment Setup. The settings below are those used by OMNIC when the performance test is run for the InGaAs detector in the main bench of the iS50 and can be used as a starting point for an experiment setup.
| Detector | InGaAs 2.6 um |
| Source | White Light |
| Beamsplitter | CaF2 |
| Optical Velocity | 0.9494 |
| Aperture | 10 |
| Attenuation | Heavy |