Thermo Fisher Scientific
Bench status report
Issue
Which FT-IR benches use the bench status report?
Environment
FT-IR
Resolution
The iS5, iS10, iS20, and iS50 Spectrometer benches use the bench status report.
  • Was this article helpful?