Czitek SurveyIR Microscope Accessory Not Recognized by eSpot Software
This article summarizes a way to resolve a problem when the eSpot software doesn't recognize that a Czitek SurveyIR microscope accessory is connected to the computer. The resolution includes using a program to update the firmware of the SurveyIR accessory.
Issue
The eSpot software provided with the Czitek SurveyIR microscope accessory does not recognize or communicate with the hardware.
Environment
Czitek SurveyIR microscope
Resolution
Use the "SurveyIR_Updater.exe" program provided by Czitek to update the accessory firmware.
Updater program is attached below and zip file includes instructions on how to update the firmware.
Analysis
The Czitek SurveyIR microscope accessory uses eSpot software for viewing the camera and controlling the microscope hardware.
This same software is also used for the Czitek MicroATR accessory.
If the eSpot software only has the option to select MicroATR, the firmware on the SurveyIR might need to be updated.
Attachment(s)
| File | Last Modified |
|---|---|
| SurveyIR Firmware Updater.zip | June 17, 2025 |