Determination if an iS10 has an Extended Baseplate Installed
Issue
Determining if a Nicolet iS10 Spectrometer has an extended baseplate installed
Environment
Nicolet iS10 Spectrometer
Resolution
The baseplate extension looks like a tongue that extends from the front of the iS10 about 3 inches and has a black aluminum plate that covers one third of the sample compartment floor area. The extension kit is required if baseplate accessories are to be used in the iS10.
The part number and description of the extension kit is:
- SAMPLE COMPARTMENT EXTENSION KIT – Optional
- 840-192100 Nicolet iS10 Sample Compartment Extension Kit
- required for large footprint accessories, standard 8"x10" baseplate mounted accessories, multi-pass gas cells
When installed the sample compartment looks like this:
Attachment(s)
| File | Last Modified |
|---|---|
| iS10_sample_compartment_installed.png | December 15, 2021 |