European Pharmacopoeia ValPro Changes
Issue
The European Pharmacopoeia (Ph. Eur.) has made changes to their guidelines on the requirements for qualifying FTIR spectrometers and ATR accessories.
Environment
FT-IR spectrometry
Resolution
Thermo Scientific ValPro software, version 2.9 reflects the changes in European Pharmacopoeia guidelines. ValPro Qualification Methods 1.1 include methods for both the FTIR spectrometer and ATR accessories.
In order to upgrade to ValPro 2.9 a customer must have OMNIC 9.8 HF2 (OMNIC 9.8.372) or higher.
For comparison, legacy and new Ph. Eur. ValPro results are included for comparison. Both the main bench and Smart iTR ATR accessory results are included as an examples.
Attachment(s)
| File | Last Modified |
|---|---|
| ValPro Qualification Test_Smart iTR Accessory - PHEUR.pdf | December 27, 2021 |
| ValPro Qualification Test_Smart iTR Accessory - LEGACY.pdf | December 27, 2021 |
| ValPro Qualification Test_Nicolet iS10 system KBr - Legacy.pdf | December 27, 2021 |
| ValPro Qualification Test_Nicolet iS10 system KBr - PHEUR.pdf | December 27, 2021 |